Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARNT
Entrez Gene ID
405
Gene Name
aryl hydrocarbon receptor nuclear translocator
Gene Aliases
HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2
Chromosome Location
Chr.1: 150809705 - 150876768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197325.1 NP_001184254.1 9 - 10 1116 70
NM_001286036.1 NP_001272965.1 10 - 11 1161 70
NM_001668.3 NP_001659.1 10 - 11 1161 70
NM_178427.2 NP_848514.1 9 - 10 1116 70
XM_005245151.1 XP_005245208.1 10 - 11 1103 70
XM_005245154.3 XP_005245211.1 11 - 12 1131 70
XM_005245157.1 XP_005245214.1 10 - 11 1103 70
XM_011509542.2 XP_011507844.1 10 - 11 1164 70
XM_011509543.2 XP_011507845.1 11 - 12 1159 70
XM_011509544.2 XP_011507846.1 10 - 11 1128 70
XM_011509545.2 XP_011507847.1 9 - 10 1119 70
XM_011509546.1 XP_011507848.1 9 - 10 879 70
XM_011509547.2 XP_011507849.1 9 - 10 1119 70
XM_017001288.1 XP_016856777.1 11 - 12 1006 70
XM_017001290.1 XP_016856779.1 10 - 11 960 70
XM_017001291.1 XP_016856780.1 10 - 11 962 70
XM_017001292.1 XP_016856781.1 11 - 12 1007 70
XM_017001293.1 XP_016856782.1 10 - 11 964 70
XM_017001294.1 XP_016856783.1 10 - 11 1103 70
XM_017001296.1 XP_016856785.1 9 - 10 1067 70
GenBank mRNA AB209877.1 - 10 - 11 972 70
AK290177.1 - 10 - 11 1100 70
AK291705.1 - 9 - 10 958 70
AK293027.1 - 10 - 11 1161 70
AK304772.1 - 9 - 10 950 70
AK313780.1 - 10 - 11 1102 70
AK315812.1 - 9 - 10 955 70
BC041121.1 - 10 - 11 975 70
BC060838.1 - 10 - 11 1096 70
M69238.1 - 10 - 11 1016 70

View Full Product Details