The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290202.1 | NP_001277131.1 | 1 - 2 | 83 | 112 |
GenBank mRNA | AK091522.1 | - | 1 - 2 | 83 | 112 |