Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGEF11
Entrez Gene ID
9826
Gene Name
Rho guanine nucleotide exchange factor 11
Gene Aliases
GTRAP48, PDZ-RHOGEF
Chromosome Location
Chr.1: 156934840 - 157046903 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014784.3 NP_055599.1 25 - 26 3403 61
NM_198236.2 NP_937879.1 26 - 27 3523 61
XM_005245629.4 XP_005245686.1 26 - 27 3927 61
XM_005245633.4 XP_005245690.1 16 - 17 1786 61
XM_006711659.2 XP_006711722.1 28 - 29 3975 61
XM_006711660.3 XP_006711723.1 28 - 29 3972 61
XM_006711661.2 XP_006711724.1 27 - 28 3945 61
XM_006711662.2 XP_006711725.1 27 - 28 3942 61
XM_006711663.3 XP_006711726.1 26 - 27 2962 61
XM_006711665.3 XP_006711728.1 27 - 28 3852 61
XM_011510185.2 XP_011508487.1 27 - 28 3960 61
XM_011510186.2 XP_011508488.1 27 - 28 3855 61
XM_011510187.2 XP_011508489.1 27 - 28 2686 61
XM_011510189.2 XP_011508491.1 5 - 6 1991 61
XM_017002920.1 XP_016858409.1 27 - 28 3942 61
XM_017002921.1 XP_016858410.1 25 - 26 3894 61
XM_017002922.1 XP_016858411.1 26 - 27 3822 61
XM_017002923.1 XP_016858412.1 25 - 26 3807 61
XM_017002924.1 XP_016858413.1 25 - 26 3807 61
GenBank mRNA AB002378.1 - 25 - 26 3108 61
AK302500.1 - 5 - 6 1000 61
BC057394.1 - 26 - 27 3547 61

View Full Product Details