Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ARHGEF11
- Entrez Gene ID
-
9826
- Gene Name
- Rho guanine nucleotide exchange factor 11
- Gene Aliases
-
GTRAP48, PDZ-RHOGEF
- Chromosome Location
-
Chr.1: 156934840 - 157046903 on Build GRCh38
View Full Product Details