Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX6
Entrez Gene ID
5190
Gene Name
peroxisomal biogenesis factor 6
Gene Aliases
HMLR2, PAF-2, PAF2, PBD4A, PDB4B, PXAAA1
Chromosome Location
Chr.6: 42963873 - 42979243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000287.3 NP_000278.3 13 - 14 2563 112
NM_001316313.1 NP_001303242.1 13 - 14 2299 112
NR_133009.1 11 - 12 2316 112
XM_011514661.1 XP_011512963.1 12 - 13 2479 112
GenBank mRNA AB051076.1 - 13 - 14 2470 112
AB051077.1 - 13 - 14 2206 112
AB051078.1 - 11 - 12 2223 112
AB208906.1 - 11 - 12 2913 112
AK314237.1 - 13 - 14 2499 112
BC033487.1 - 13 - 14 2478 112
BC048331.1 - 13 - 14 2486 112
D83703.1 - 13 - 14 2540 112

View Full Product Details