The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000287.3 | NP_000278.3 | 2 - 3 | 1139 | 93 |
NM_001316313.1 | NP_001303242.1 | 2 - 3 | 875 | 93 | |
NR_133009.1 | 2 - 3 | 1139 | 93 | ||
GenBank mRNA | AB051076.1 | - | 2 - 3 | 1046 | 93 |
AB051077.1 | - | 2 - 3 | 782 | 93 | |
AB051078.1 | - | 2 - 3 | 1046 | 93 | |
AB208906.1 | - | 2 - 3 | 1099 | 93 | |
AK225704.1 | - | 2 - 3 | 1077 | 93 | |
AK314237.1 | - | 2 - 3 | 1075 | 93 | |
BC033487.1 | - | 2 - 3 | 1054 | 93 | |
BC048331.1 | - | 2 - 3 | 1062 | 93 |