Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX6
Entrez Gene ID
5190
Gene Name
peroxisomal biogenesis factor 6
Gene Aliases
HMLR2, PAF-2, PAF2, PBD4A, PDB4B, PXAAA1
Chromosome Location
Chr.6: 42963873 - 42979243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000287.3 NP_000278.3 8 - 9 1973 60
NM_001316313.1 NP_001303242.1 8 - 9 1709 60
NR_133009.1 8 - 9 1973 60
XM_011514661.1 XP_011512963.1 7 - 8 1889 60
GenBank mRNA AB051076.1 - 8 - 9 1880 60
AB051077.1 - 8 - 9 1616 60
AB051078.1 - 8 - 9 1880 60
AB208906.1 - 8 - 9 1933 60
AK314237.1 - 8 - 9 1909 60
BC033487.1 - 8 - 9 1888 60
BC048331.1 - 8 - 9 1896 60
D83703.1 - 8 - 9 1950 60

View Full Product Details