The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015384.4 | NP_056199.2 | 44 - 45 | 8178 | 116 |
NM_133433.3 | NP_597677.2 | 44 - 45 | 8178 | 116 | |
XM_005248280.3 | XP_005248337.1 | 44 - 45 | 8167 | 116 | |
XM_005248282.4 | XP_005248339.3 | 38 - 39 | 7669 | 116 | |
XM_006714467.2 | XP_006714530.1 | 43 - 44 | 8020 | 116 | |
XM_006714468.2 | XP_006714531.1 | 43 - 44 | 7969 | 116 | |
XM_017009329.1 | XP_016864818.1 | 43 - 44 | 8020 | 116 | |
XM_017009330.1 | XP_016864819.1 | 36 - 37 | 10441 | 116 | |
XM_017009331.1 | XP_016864820.1 | 43 - 44 | 6541 | 116 | |
GenBank mRNA | AJ627032.1 | - | 44 - 45 | 8147 | 116 |
AJ640137.1 | - | 44 - 45 | 8147 | 116 | |
AK002065.1 | - | 8 - 9 | 1296 | 116 | |
AK022659.1 | - | 10 - 11 | 1490 | 116 | |
AK023416.1 | - | 12 - 13 | 1826 | 116 | |
AL137726.1 | - | 3 - 4 | 443 | 116 | |
BC033847.1 | - | 3 - 4 | 460 | 116 |