The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321635.1 | NP_001308564.1 | 8 - 9 | 1131 | 89 |
NM_001321636.1 | NP_001308565.1 | 8 - 9 | 1131 | 89 | |
NM_024759.2 | NP_079035.1 | 8 - 9 | 1131 | 89 | |
NR_135745.1 | 9 - 10 | 1295 | 89 | ||
NR_135746.1 | 10 - 11 | 1417 | 89 | ||
NR_135747.1 | 9 - 10 | 1355 | 89 | ||
GenBank mRNA | AK024017.1 | - | 8 - 9 | 1131 | 89 |
AK302414.1 | - | 7 - 8 | 894 | 89 | |
BC132687.1 | - | 8 - 9 | 1074 | 89 | |
BC144055.1 | - | 8 - 9 | 1074 | 89 |