The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135699.1 | NP_001129171.1 | 5 - 6 | 836 | 81 |
NM_001135700.1 | NP_001129172.1 | 5 - 6 | 790 | 81 | |
NM_001135701.1 | NP_001129173.1 | 5 - 6 | 839 | 81 | |
NM_001135702.1 | NP_001129174.1 | 5 - 6 | 858 | 81 | |
NM_003406.3 | NP_003397.1 | 5 - 6 | 819 | 81 | |
NM_145690.2 | NP_663723.1 | 5 - 6 | 893 | 81 | |
XM_005251061.3 | XP_005251118.1 | 7 - 8 | 1216 | 81 | |
XM_005251063.3 | XP_005251120.1 | 5 - 6 | 988 | 81 | |
XM_017013810.1 | XP_016869299.1 | 6 - 7 | 1005 | 81 | |
XM_017013811.1 | XP_016869300.1 | 7 - 8 | 1201 | 81 | |
GenBank mRNA | AK289945.1 | - | 5 - 6 | 729 | 81 |
AK294647.1 | - | 4 - 5 | 478 | 81 | |
AK296902.1 | - | 4 - 5 | 473 | 81 | |
AK307506.1 | - | 6 - 7 | 696 | 81 | |
AK315867.1 | - | 5 - 6 | 607 | 81 | |
BC003623.2 | - | 5 - 6 | 786 | 81 | |
BC013265.1 | - | 5 - 6 | 784 | 81 | |
BC051814.1 | - | 5 - 6 | 907 | 81 | |
BC063824.1 | - | 5 - 6 | 758 | 81 | |
BC068456.1 | - | 5 - 6 | 807 | 81 | |
BC072426.1 | - | 5 - 6 | 788 | 81 | |
BC073141.1 | - | 5 - 6 | 742 | 81 | |
BC083508.1 | - | 5 - 6 | 786 | 81 | |
BC099904.1 | - | 5 - 6 | 808 | 81 | |
BC101483.1 | - | 5 - 6 | 741 | 81 | |
BC108281.1 | - | 5 - 6 | 796 | 81 | |
BC111951.1 | - | 5 - 6 | 741 | 81 | |
DQ185048.1 | - | 2 - 3 | 265 | 81 | |
EU794582.1 | - | 5 - 6 | 787 | 81 | |
EU794583.1 | - | 5 - 6 | 807 | 81 | |
GQ472215.1 | - | 5 - 6 | 807 | 81 | |
M86400.1 | - | 5 - 6 | 765 | 81 | |
U28964.1 | - | 5 - 6 | 807 | 81 |