The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001315529.1 | NP_001302458.1 | 36 - 37 | 8180 | 102 |
NM_006031.5 | NP_006022.3 | 36 - 37 | 8018 | 102 | |
XM_005261124.4 | XP_005261181.1 | 37 - 38 | 8070 | 102 | |
XM_011529594.2 | XP_011527896.1 | 37 - 38 | 8118 | 102 | |
XM_017028362.1 | XP_016883851.1 | 36 - 37 | 8037 | 102 | |
XM_017028363.1 | XP_016883852.1 | 37 - 38 | 8239 | 102 | |
GenBank mRNA | AB007862.3 | - | 36 - 37 | 8181 | 102 |
AF515282.1 | - | 36 - 37 | 7911 | 102 | |
U52962.1 | - | 37 - 38 | 7930 | 102 |