Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPNE3
Entrez Gene ID
8895
Gene Name
copine 3
Gene Aliases
CPN3, PRO1071
Chromosome Location
Chr.8: 86514423 - 86561498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003909.4 NP_003900.1 16 - 17 1651 69
XM_005251093.4 XP_005251150.1 15 - 16 1603 69
XM_017013945.1 XP_016869434.1 17 - 18 1776 69
GenBank mRNA AB014536.1 - 16 - 17 1608 69
AF077226.2 - 15 - 16 1549 69
AK292912.1 - 16 - 17 1640 69
BC036242.1 - 16 - 17 1611 69
BC066597.1 - 16 - 17 1622 69
BX647342.1 - 6 - 7 1074 69

View Full Product Details