Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPNE3
Entrez Gene ID
8895
Gene Name
copine 3
Gene Aliases
CPN3, PRO1071
Chromosome Location
Chr.8: 86514423 - 86561498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003909.4 NP_003900.1 8 - 9 795 81
XM_005251093.4 XP_005251150.1 7 - 8 747 81
XM_017013945.1 XP_016869434.1 9 - 10 920 81
GenBank mRNA AB014536.1 - 8 - 9 752 81
AF077226.2 - 7 - 8 693 81
AK292912.1 - 8 - 9 784 81
BC007017.1 - 8 - 9 779 81
BC008385.1 - 8 - 9 779 81
BC036242.1 - 8 - 9 755 81
BC066597.1 - 8 - 9 766 81

View Full Product Details