The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000168.5 | NP_000159.3 | 10 - 11 | 1585 | 76 |
XM_011515274.2 | XP_011513576.1 | 9 - 10 | 4209 | 76 | |
XM_017011997.1 | XP_016867486.1 | 9 - 10 | 4242 | 76 | |
GenBank mRNA | AK299299.1 | - | 9 - 10 | 1471 | 76 |
AK308429.1 | - | 11 - 12 | 1650 | 76 | |
BC113616.1 | - | 9 - 10 | 1542 | 76 | |
BC117168.1 | - | 9 - 10 | 1542 | 76 | |
M57609.1 | - | 9 - 10 | 1548 | 76 |