The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015474.3 | NP_056289.2 | 12 - 13 | 1609 | 71 |
XM_005260384.3 | XP_005260441.1 | 12 - 13 | 1582 | 71 | |
XM_011528761.2 | XP_011527063.1 | 12 - 13 | 1582 | 71 | |
GenBank mRNA | AB013847.1 | - | 12 - 13 | 1409 | 71 |
AB208944.1 | - | 12 - 13 | 1434 | 71 | |
AF228421.1 | - | 12 - 13 | 1562 | 71 | |
AK027811.1 | - | 12 - 13 | 1582 | 71 | |
AK304187.1 | - | 12 - 13 | 1366 | 71 | |
AK311150.1 | - | 1 - 2 | 44 | 71 | |
AK315169.1 | - | 12 - 13 | 1542 | 71 | |
AL050267.1 | - | 12 - 13 | 1484 | 71 | |
BC036450.2 | - | 12 - 13 | 1437 | 71 |