The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002657.3 | NP_002648.1 | 2 - 3 | 478 | 114 |
XM_005260436.3 | XP_005260493.1 | 2 - 3 | 834 | 114 | |
XM_011528863.2 | XP_011527165.1 | 2 - 3 | 1323 | 114 | |
XM_011528864.2 | XP_011527166.1 | 2 - 3 | 915 | 114 | |
GenBank mRNA | AF006005.1 | - | 2 - 3 | 463 | 114 |
AK292450.1 | - | 2 - 3 | 446 | 114 | |
BC023655.2 | - | 2 - 3 | 464 | 114 | |
D83784.1 | - | 2 - 3 | 461 | 114 |