The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004798.3 | NP_004789.1 | 6 - 7 | 2037 | 157 |
GenBank mRNA | AB002357.1 | - | 6 - 7 | 2024 | 157 |
AK098431.1 | - | 5 - 6 | 582 | 157 | |
AK299877.1 | - | 6 - 7 | 960 | 157 | |
BC136310.1 | - | 5 - 6 | 1930 | 157 | |
BC136311.1 | - | 5 - 6 | 1930 | 157 |