The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014819.4 | NP_055634.3 | 4 - 5 | 1506 | 83 |
XM_017010099.1 | XP_016865588.1 | 5 - 6 | 1582 | 83 | |
GenBank mRNA | AB007898.1 | - | 4 - 5 | 1403 | 83 |
AK291759.1 | - | 4 - 5 | 1440 | 83 | |
BC030826.1 | - | 4 - 5 | 1402 | 83 | |
BP301854.1 | - | 1 - 2 | 443 | 83 |