The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207012.1 | NP_001193941.1 | 5 - 6 | 950 | 157 |
GenBank mRNA | A18411.1 | - | 5 - 6 | 749 | 157 |
X54936.1 | - | 5 - 6 | 749 | 157 |