The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207012.1 | NP_001193941.1 | 4 - 5 | 917 | 73 |
GenBank mRNA | A18411.1 | - | 4 - 5 | 716 | 73 |
X54936.1 | - | 4 - 5 | 716 | 73 |