Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AK2
Entrez Gene ID
204
Gene Name
adenylate kinase 2
Gene Aliases
ADK2
Chromosome Location
Chr.1: 33007940 - 33036911 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199199.1 NP_001186128.1 2 - 3 301 68
NM_001319139.1 NP_001306068.1 3 - 4 419 68
NM_001319140.1 NP_001306069.1 3 - 4 419 68
NM_001319141.1 NP_001306070.1 2 - 3 301 68
NM_001319143.1 NP_001306072.1 2 - 3 301 68
NM_001625.3 NP_001616.1 2 - 3 301 68
NM_013411.4 NP_037543.1 2 - 3 301 68
NR_134976.1 2 - 3 301 68
GenBank mRNA AB005621.1 - 2 - 3 256 68
AB005622.1 - 2 - 3 260 68
AB451267.1 - 2 - 3 218 68
AB451394.1 - 2 - 3 218 68
AK291676.1 - 2 - 3 238 68
AK295105.1 - 3 - 4 352 68
AK296863.1 - 2 - 3 235 68
AK297036.1 - 2 - 3 238 68
AL557311.3 - 2 - 3 275 68
AY080899.1 - 2 - 3 253 68
AY080900.1 - 3 - 4 339 68
BC009405.2 - 2 - 3 229 68
BC026705.1 - 2 - 3 237 68
BC070127.1 - 2 - 3 243 68
BC090040.1 - 2 - 3 243 68
BQ889289.1 - 3 - 4 385 68
BU625169.1 - 3 - 4 68
BX504559.1 - 2 - 3 282 68
DC422465.1 - 2 - 3 301 68
DN601721.1 - 2 - 3 220 68
HY061267.1 - 2 - 3 302 68
U39945.1 - 2 - 3 251 68
U54645.1 - 2 - 3 221 68
U84371.1 - 2 - 3 249 68

View Full Product Details