Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AK2
Entrez Gene ID
204
Gene Name
adenylate kinase 2
Gene Aliases
ADK2
Chromosome Location
Chr.1: 33007940 - 33036911 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319141.1 NP_001306070.1 7 - 8 820 73
GenBank mRNA AL557311.3 - 7 - 8 793 73

View Full Product Details