Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTN5
Entrez Gene ID
53942
Gene Name
contactin 5
Gene Aliases
HNB-2s, NB-2
Chromosome Location
Chr.11: 99020904 - 100358885 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243270.1 NP_001230199.1 2 - 3 139
NM_001243271.1 NP_001230200.1 3 - 4 139
NM_014361.3 NP_055176.1 3 - 4 139
XM_011542873.1 XP_011541175.1 3 - 4 139
XM_017017926.1 XP_016873415.1 4 - 5 139
XM_017017927.1 XP_016873416.1 4 - 5 139
XM_017017928.1 XP_016873417.1 2 - 3 139
GenBank mRNA AB013802.2 - 1 - 2 139
AK308788.1 - 3 - 4 139
BC039255.1 - 3 - 4 388 139
BC130619.1 - 2 - 3 139
BC144185.1 - 2 - 3 139
BC144186.1 - 3 - 4 139

View Full Product Details