The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243565.1 | NP_001230494.1 | 4 - 5 | 478 | 62 |
GenBank mRNA | BC004117.2 | - | 4 - 5 | 420 | 62 |