The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017589.3 | NP_060059.1 | 2 - 3 | 373 | 102 |
XM_011542876.2 | XP_011541178.1 | 2 - 3 | 407 | 102 | |
XM_011542878.2 | XP_011541180.1 | 2 - 3 | 407 | 102 | |
XM_011542879.2 | XP_011541181.1 | 2 - 3 | 407 | 102 | |
XM_011542880.2 | XP_011541182.1 | 2 - 3 | 407 | 102 | |
GenBank mRNA | AJ271351.1 | - | 1 - 2 | 178 | 102 |
BC031045.1 | - | 2 - 3 | 359 | 102 | |
BX099301.1 | - | 2 - 3 | 373 | 102 |