Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
Chromosome Location
Chr.1: 33513999 - 34165842 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281956.1 NP_001268885.1 16 - 17 2683 71
NM_052896.4 NP_443128.2 16 - 17 2415 71
XM_017000185.1 XP_016855674.1 16 - 17 2677 71
XM_017000186.1 XP_016855675.1 16 - 17 2677 71
XM_017000187.1 XP_016855676.1 16 - 17 2506 71
XM_017000188.1 XP_016855677.1 16 - 17 2415 71
XM_017000189.1 XP_016855678.1 16 - 17 2677 71
XM_017000190.1 XP_016855679.1 10 - 11 1942 71
XM_017000191.1 XP_016855680.1 16 - 17 2677 71
XM_017000192.1 XP_016855681.1 16 - 17 2677 71
XM_017000193.1 XP_016855682.1 16 - 17 2677 71
XM_017000194.1 XP_016855683.1 16 - 17 2415 71
GenBank mRNA AB212622.1 - 16 - 17 2683 71
AY210418.1 - 16 - 17 2415 71

View Full Product Details