Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIFR
Entrez Gene ID
3977
Gene Name
leukemia inhibitory factor receptor alpha
Gene Aliases
CD118, LIF-R, SJS2, STWS, SWS
Chromosome Location
Chr.5: 38474963 - 38606290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127671.1 NP_001121143.1 15 - 16 2499 118
NM_002310.5 NP_002301.1 15 - 16 2331 118
XM_011514040.2 XP_011512342.1 15 - 16 2517 118
XM_011514042.2 XP_011512344.1 16 - 17 2553 118
XM_017009462.1 XP_016864951.1 15 - 16 2344 118
XM_017009463.1 XP_016864952.1 16 - 17 2395 118
GenBank mRNA AK290059.1 - 15 - 16 2499 118
AK308158.1 - 15 - 16 2330 118
X61615.1 - 15 - 16 2345 118

View Full Product Details