The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002372.3 | NP_002363.2 | 20 - 21 | 4226 | 83 |
XM_017009472.1 | XP_016864961.1 | 20 - 21 | 3147 | 83 | |
GenBank mRNA | AK314090.1 | - | 20 - 21 | 3567 | 83 |
BC043416.1 | - | 1 - 2 | 1761 | 83 | |
BC142656.1 | - | 20 - 21 | 3525 | 83 | |
BC142696.1 | - | 20 - 21 | 3525 | 83 | |
D63998.1 | - | 20 - 21 | 3681 | 83 | |
U31520.1 | - | 20 - 21 | 3177 | 83 |