The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002372.3 | NP_002363.2 | 6 - 7 | 2063 | 93 |
XM_011543395.2 | XP_011541697.1 | 6 - 7 | 1502 | 93 | |
XM_017009472.1 | XP_016864961.1 | 6 - 7 | 984 | 93 | |
GenBank mRNA | AK314090.1 | - | 6 - 7 | 1404 | 93 |
BC142656.1 | - | 6 - 7 | 1365 | 93 | |
BC142696.1 | - | 6 - 7 | 1365 | 93 | |
D63998.1 | - | 6 - 7 | 1521 | 93 | |
U31520.1 | - | 6 - 7 | 1014 | 93 |