The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014598.3 | NP_055413.1 | 2 - 3 | 975 | 74 |
XM_017024551.1 | XP_016880040.1 | 2 - 3 | 1223 | 74 | |
GenBank mRNA | AB005216.1 | - | 2 - 3 | 486 | 74 |
BC121030.1 | - | 3 - 4 | 558 | 74 | |
BC121031.1 | - | 3 - 4 | 507 | 74 | |
BC128607.1 | - | 2 - 3 | 431 | 74 |