The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014598.3 | NP_055413.1 | 8 - 9 | 1744 | 121 |
XM_017024551.1 | XP_016880040.1 | 7 - 8 | 1890 | 121 | |
XM_017024552.1 | XP_016880041.1 | 6 - 7 | 1785 | 121 | |
GenBank mRNA | AA233668.1 | - | 2 - 3 | 184 | 121 |
AB005216.1 | - | 8 - 9 | 1255 | 121 | |
AI223248.1 | - | 2 - 3 | 121 | ||
BC121031.1 | - | 8 - 9 | 1174 | 121 | |
BC128607.1 | - | 8 - 9 | 1200 | 121 |