Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIN1
Entrez Gene ID
54826
Gene Name
gypsy retrotransposon integrase 1
Gene Aliases
GIN-1, TGIN1, ZH2C2
Chromosome Location
Chr.5: 103086000 - 103120149 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317954.1 NP_001304883.1 5 - 6 897 157
NM_017676.2 NP_060146.2 6 - 7 1091 157
XM_005272028.3 XP_005272085.1 6 - 7 1098 157
XM_017009599.1 XP_016865088.1 6 - 7 2526 157
GenBank mRNA AK000132.1 - 6 - 7 1089 157
AK225572.1 - 6 - 7 1088 157
AK295796.1 - 6 - 7 1079 157
BC151135.1 - 5 - 6 1031 157
BC157835.1 - 5 - 6 1031 157

View Full Product Details