Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF362
Entrez Gene ID
149076
Gene Name
zinc finger protein 362
Gene Aliases
RN, lin-29
Chromosome Location
Chr.1: 33256307 - 33300719 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152493.2 NP_689706.2 6 - 7 1079 80
XM_005270507.2 XP_005270564.1 5 - 6 1219 80
XM_005270508.4 XP_005270565.1 6 - 7 1876 80
XM_017000415.1 XP_016855904.1 5 - 6 1791 80
GenBank mRNA AF318356.1 - 5 - 6 929 80
BC104779.1 - 5 - 6 1001 80
BC112113.1 - 5 - 6 1001 80

View Full Product Details