Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF19B
Entrez Gene ID
127544
Gene Name
ring finger protein 19B
Gene Aliases
IBRDC3, NKLAM
Chromosome Location
Chr.1: 32932498 - 32965266 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127361.2 NP_001120833.1 2 - 3 969 109
NM_001300826.1 NP_001287755.1 2 - 3 969 109
GenBank mRNA AK131439.1 - 3 - 4 666 109
BC062374.1 - 2 - 3 446 109
BC143687.1 - 2 - 3 336 109
DA630289.1 - 3 - 4 481 109

View Full Product Details