The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127361.2 | NP_001120833.1 | 4 - 5 | 1280 | 94 |
NM_001300826.1 | NP_001287755.1 | 4 - 5 | 1280 | 94 | |
NM_153341.3 | NP_699172.2 | 4 - 5 | 1283 | 94 | |
XM_006710356.2 | XP_006710419.1 | 4 - 5 | 1736 | 94 | |
XM_006710357.3 | XP_006710420.1 | 4 - 5 | 1736 | 94 | |
XM_006710358.3 | XP_006710421.1 | 4 - 5 | 1736 | 94 | |
GenBank mRNA | AK074486.1 | - | 4 - 5 | 583 | 94 |
AK131439.1 | - | 5 - 6 | 977 | 94 | |
BC062374.1 | - | 4 - 5 | 757 | 94 | |
BC113560.1 | - | 4 - 5 | 650 | 94 | |
BC143687.1 | - | 4 - 5 | 647 | 94 |