Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF19B
Entrez Gene ID
127544
Gene Name
ring finger protein 19B
Gene Aliases
IBRDC3, NKLAM
Chromosome Location
Chr.1: 32932498 - 32965266 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127361.2 NP_001120833.1 5 - 6 1388 97
NM_001300826.1 NP_001287755.1 5 - 6 1388 97
NM_153341.3 NP_699172.2 5 - 6 1391 97
XM_006710356.2 XP_006710419.1 5 - 6 1844 97
XM_006710357.3 XP_006710420.1 5 - 6 1844 97
XM_006710358.3 XP_006710421.1 5 - 6 1844 97
GenBank mRNA AK074486.1 - 5 - 6 691 97
AK131439.1 - 6 - 7 1085 97
BC020595.2 - 1 - 2 96 97
BC062374.1 - 5 - 6 865 97
BC113560.1 - 5 - 6 758 97
BC143687.1 - 5 - 6 755 97

View Full Product Details