The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002889.3 | NP_002880.1 | 1 - 2 | 99 | 89 |
XM_017012491.1 | XP_016867980.1 | 1 - 2 | 110 | 89 | |
GenBank mRNA | BX448753.2 | - | 1 - 2 | 99 | 89 |