The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015347.4 | NP_056162.4 | 8 - 9 | 1617 | 72 |
XM_011538102.1 | XP_011536404.1 | 9 - 10 | 1718 | 72 | |
XM_011538103.2 | XP_011536405.1 | 11 - 12 | 2180 | 72 | |
XM_011538104.2 | XP_011536406.1 | 11 - 12 | 1995 | 72 | |
XM_011538105.2 | XP_011536407.1 | 11 - 12 | 2169 | 72 | |
XM_011538106.2 | XP_011536408.1 | 11 - 12 | 2169 | 72 | |
XM_011538107.2 | XP_011536409.1 | 11 - 12 | 2176 | 72 | |
XM_011538108.2 | XP_011536410.1 | 11 - 12 | 2175 | 72 | |
XM_017019105.1 | XP_016874594.1 | 11 - 12 | 2193 | 72 | |
XM_017019106.1 | XP_016874595.1 | 11 - 12 | 2169 | 72 | |
XM_017019107.1 | XP_016874596.1 | 11 - 12 | 1991 | 72 | |
XM_017019108.1 | XP_016874597.1 | 11 - 12 | 2174 | 72 | |
XM_017019109.1 | XP_016874598.1 | 9 - 10 | 1718 | 72 | |
XM_017019110.1 | XP_016874599.1 | 11 - 12 | 1991 | 72 | |
XM_017019111.1 | XP_016874600.1 | 11 - 12 | 1991 | 72 | |
XM_017019112.1 | XP_016874601.1 | 11 - 12 | 2213 | 72 | |
GenBank mRNA | AB002316.1 | - | 8 - 9 | 1617 | 72 |
AY699979.1 | - | 5 - 6 | 1308 | 72 | |
BC007632.1 | - | 12 - 13 | 2504 | 72 |