Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC51A
Entrez Gene ID
200931
Gene Name
solute carrier family 51 alpha subunit
Gene Aliases
OSTA, OSTalpha
Chromosome Location
Chr.3: 196216512 - 196233430 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152672.5 NP_689885.4 1 - 2 238 63
GenBank mRNA AK172837.1 - 1 - 2 241 63
AK300996.1 - 1 - 2 236 63
AY194243.1 - 1 - 2 59 63

View Full Product Details