The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000102.3 | NP_000093.1 | 5 - 6 | 1143 | 65 |
GenBank mRNA | AK289898.1 | - | 5 - 6 | 1023 | 65 |
AK308466.1 | - | 5 - 6 | 933 | 65 | |
AK312235.1 | - | 5 - 6 | 1025 | 65 | |
BC062997.1 | - | 5 - 6 | 995 | 65 | |
BC063388.1 | - | 5 - 6 | 995 | 65 | |
BT020000.1 | - | 5 - 6 | 971 | 65 | |
M14564.1 | - | 5 - 6 | 1012 | 65 |