The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178858.4 | NP_849189.1 | 4 - 5 | 601 | 78 |
XM_011539261.2 | XP_011537563.1 | 5 - 6 | 602 | 78 | |
XM_011539262.2 | XP_011537564.1 | 5 - 6 | 722 | 78 | |
XM_011539263.2 | XP_011537565.1 | 6 - 7 | 766 | 78 | |
XM_011539264.2 | XP_011537566.1 | 5 - 6 | 628 | 78 | |
GenBank mRNA | AF462052.1 | - | 4 - 5 | 556 | 78 |
AK308246.1 | - | 4 - 5 | 506 | 78 | |
BC022091.2 | - | 4 - 5 | 601 | 78 |