Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP3
Entrez Gene ID
10566
Gene Name
A-kinase anchoring protein 3
Gene Aliases
AKAP 110, AKAP110, CT82, FSP95, HEL159, PRKA3, SOB1
Chromosome Location
Chr.12: 4615508 - 4649047 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278309.1 NP_001265238.1 3 - 4 552 124
NM_006422.3 NP_006413.3 3 - 4 530 124
XM_005253664.2 XP_005253721.1 3 - 4 462 124
XM_011520908.1 XP_011519210.1 2 - 3 666 124
GenBank mRNA AF093408.1 - 2 - 3 234 124
AK292451.1 - 3 - 4 530 124
BC047535.1 - 2 - 3 214 124
EU794681.1 - 2 - 3 234 124
U85715.1 - 2 - 3 207 124

View Full Product Details