The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278309.1 | NP_001265238.1 | 5 - 6 | 2951 | 109 |
NM_006422.3 | NP_006413.3 | 5 - 6 | 2929 | 109 | |
XM_005253664.2 | XP_005253721.1 | 5 - 6 | 2861 | 109 | |
XM_011520908.1 | XP_011519210.1 | 4 - 5 | 3065 | 109 | |
XM_011520909.1 | XP_011519211.1 | 3 - 4 | 2542 | 109 | |
XM_011520910.2 | XP_011519212.1 | 3 - 4 | 2997 | 109 | |
GenBank mRNA | AF087003.1 | - | 4 - 5 | 2566 | 109 |
AF093408.1 | - | 4 - 5 | 2633 | 109 | |
AK074722.1 | - | 1 - 2 | 2022 | 109 | |
AK292451.1 | - | 5 - 6 | 2929 | 109 | |
BC047535.1 | - | 4 - 5 | 2613 | 109 | |
EU794681.1 | - | 4 - 5 | 2633 | 109 | |
U85715.1 | - | 4 - 5 | 2606 | 109 |