The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005736.3 | NP_005727.1 | 5 - 6 | 554 | 84 |
GenBank mRNA | AK297620.1 | - | 4 - 5 | 448 | 84 |
AK302072.1 | - | 4 - 5 | 443 | 84 | |
AK312506.1 | - | 5 - 6 | 513 | 84 | |
BC000693.2 | - | 5 - 6 | 490 | 84 | |
BC026016.2 | - | 5 - 6 | 456 | 84 | |
X82206.1 | - | 5 - 6 | 512 | 84 | |
Z14978.1 | - | 5 - 6 | 494 | 84 |