Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTTN
Entrez Gene ID
2017
Gene Name
cortactin
Gene Aliases
EMS1
Chromosome Location
Chr.11: 70398506 - 70436584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184740.1 NP_001171669.1 6 - 7 606 68
NM_005231.3 NP_005222.2 6 - 7 606 68
NM_138565.2 NP_612632.1 6 - 7 606 68
XM_006718447.2 XP_006718510.1 6 - 7 606 68
XM_006718448.3 XP_006718511.1 6 - 7 606 68
XM_017017312.1 XP_016872801.1 6 - 7 606 68
GenBank mRNA AK222613.1 - 6 - 7 575 68
AK291097.1 - 6 - 7 577 68
AU124856.1 - 6 - 7 574 68
BC008799.2 - 6 - 7 568 68
BC033889.1 - 6 - 7 576 68
M98343.1 - 6 - 7 568 68

View Full Product Details