Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP42
Entrez Gene ID
132625
Gene Name
ZFP42 zinc finger protein
Gene Aliases
REX-1, REX1, ZNF754, zfp-42
Chromosome Location
Chr.4: 187994044 - 188005050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011531604.2 XP_011529906.1 2 - 3 165 105
XM_011531605.1 XP_011529907.1 3 - 4 315 105
XM_011531606.1 XP_011529908.1 3 - 4 319 105
XM_011531607.2 XP_011529909.1 2 - 3 162 105

View Full Product Details