Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRM
Entrez Gene ID
5797
Gene Name
protein tyrosine phosphatase, receptor type M
Gene Aliases
PTPRL1, R-PTP-MU, RPTPM, RPTPU, hR-PTPu
Chromosome Location
Chr.18: 7567316 - 8406861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105244.1 NP_001098714.1 14 - 15 2803 140
NM_002845.3 NP_002836.3 14 - 15 2803 140
XM_011525708.2 XP_011524010.1 15 - 16 2873 140
XM_011525710.2 XP_011524012.1 15 - 16 2873 140
XM_011525712.2 XP_011524014.1 14 - 15 2798 140
XM_011525713.2 XP_011524015.1 15 - 16 2473 140
XM_011525715.2 XP_011524017.1 11 - 12 1838 140
XM_011525716.2 XP_011524018.1 10 - 11 1757 140
XM_011525717.2 XP_011524019.1 9 - 10 1547 140
XM_011525722.2 XP_011524024.1 10 - 11 1763 140
XM_017025894.1 XP_016881383.1 15 - 16 2417 140
XM_017025895.1 XP_016881384.1 15 - 16 2417 140
XM_017025896.1 XP_016881385.1 15 - 16 2417 140
XM_017025897.1 XP_016881386.1 15 - 16 2417 140
XM_017025898.1 XP_016881387.1 15 - 16 2417 140
XM_017025899.1 XP_016881388.1 14 - 15 2342 140
XM_017025900.1 XP_016881389.1 15 - 16 2417 140
XM_017025901.1 XP_016881390.1 14 - 15 2342 140
XM_017025902.1 XP_016881391.1 14 - 15 2342 140
XM_017025903.1 XP_016881392.1 14 - 15 2342 140
XM_017025904.1 XP_016881393.1 14 - 15 2342 140
XM_017025905.1 XP_016881394.1 15 - 16 2473 140
XM_017025906.1 XP_016881395.1 11 - 12 1822 140
XM_017025907.1 XP_016881396.1 10 - 11 1763 140
XM_017025908.1 XP_016881397.1 11 - 12 1838 140
XM_017025909.1 XP_016881398.1 10 - 11 1763 140
GenBank mRNA AK294492.1 - 10 - 11 1684 140
AK303652.1 - 14 - 15 2301 140
BC040543.1 - 2 - 3 597 140
BC051651.1 - 14 - 15 2539 140
BC151842.1 - 14 - 15 3349 140
X58288.1 - 14 - 15 2300 140

View Full Product Details