Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRM
Entrez Gene ID
5797
Gene Name
protein tyrosine phosphatase, receptor type M
Gene Aliases
PTPRL1, R-PTP-MU, RPTPM, RPTPU, hR-PTPu
Chromosome Location
Chr.18: 7567316 - 8406861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105244.1 NP_001098714.1 27 - 28 4112 86
NM_002845.3 NP_002836.3 25 - 26 4073 86
XM_011525708.2 XP_011524010.1 29 - 30 4233 86
XM_011525710.2 XP_011524012.1 28 - 29 4182 86
XM_011525712.2 XP_011524014.1 28 - 29 4158 86
XM_011525713.2 XP_011524015.1 29 - 30 3833 86
XM_011525715.2 XP_011524017.1 25 - 26 3198 86
XM_011525716.2 XP_011524018.1 21 - 22 3027 86
XM_011525717.2 XP_011524019.1 23 - 24 2907 86
XM_011525722.2 XP_011524024.1 23 - 24 3072 86
XM_017025894.1 XP_016881383.1 29 - 30 3777 86
XM_017025895.1 XP_016881384.1 28 - 29 3765 86
XM_017025896.1 XP_016881385.1 27 - 28 3738 86
XM_017025897.1 XP_016881386.1 28 - 29 3726 86
XM_017025898.1 XP_016881387.1 27 - 28 3714 86
XM_017025899.1 XP_016881388.1 28 - 29 3702 86
XM_017025900.1 XP_016881389.1 26 - 27 3687 86
XM_017025901.1 XP_016881390.1 26 - 27 3663 86
XM_017025902.1 XP_016881391.1 27 - 28 3651 86
XM_017025903.1 XP_016881392.1 26 - 27 3639 86
XM_017025904.1 XP_016881393.1 25 - 26 3612 86
XM_017025905.1 XP_016881394.1 28 - 29 3782 86
XM_017025906.1 XP_016881395.1 24 - 25 3131 86
XM_017025907.1 XP_016881396.1 24 - 25 3123 86
XM_017025908.1 XP_016881397.1 22 - 23 3108 86
XM_017025909.1 XP_016881398.1 22 - 23 3060 86
GenBank mRNA AK294492.1 - 21 - 22 2954 86
BC040543.1 - 11 - 12 3346 86
BC051651.1 - 25 - 26 3809 86
BC151842.1 - 27 - 28 4658 86
X58288.1 - 25 - 26 3570 86

View Full Product Details