Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRM
Entrez Gene ID
5797
Gene Name
protein tyrosine phosphatase, receptor type M
Gene Aliases
PTPRL1, R-PTP-MU, RPTPM, RPTPU, hR-PTPu
Chromosome Location
Chr.18: 7567316 - 8406861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105244.1 NP_001098714.1 28 - 29 4290 74
NM_002845.3 NP_002836.3 26 - 27 4251 74
XM_011525708.2 XP_011524010.1 30 - 31 4411 74
XM_011525710.2 XP_011524012.1 29 - 30 4360 74
XM_011525712.2 XP_011524014.1 29 - 30 4336 74
XM_011525713.2 XP_011524015.1 30 - 31 4011 74
XM_011525715.2 XP_011524017.1 26 - 27 3376 74
XM_011525716.2 XP_011524018.1 22 - 23 3205 74
XM_011525717.2 XP_011524019.1 24 - 25 3085 74
XM_011525722.2 XP_011524024.1 24 - 25 3250 74
XM_017025894.1 XP_016881383.1 30 - 31 3955 74
XM_017025895.1 XP_016881384.1 29 - 30 3943 74
XM_017025896.1 XP_016881385.1 28 - 29 3916 74
XM_017025897.1 XP_016881386.1 29 - 30 3904 74
XM_017025898.1 XP_016881387.1 28 - 29 3892 74
XM_017025899.1 XP_016881388.1 29 - 30 3880 74
XM_017025900.1 XP_016881389.1 27 - 28 3865 74
XM_017025901.1 XP_016881390.1 27 - 28 3841 74
XM_017025902.1 XP_016881391.1 28 - 29 3829 74
XM_017025903.1 XP_016881392.1 27 - 28 3817 74
XM_017025904.1 XP_016881393.1 26 - 27 3790 74
XM_017025905.1 XP_016881394.1 29 - 30 3960 74
XM_017025906.1 XP_016881395.1 25 - 26 3309 74
XM_017025907.1 XP_016881396.1 25 - 26 3301 74
XM_017025908.1 XP_016881397.1 23 - 24 3286 74
XM_017025909.1 XP_016881398.1 23 - 24 3238 74
GenBank mRNA AK294492.1 - 22 - 23 3132 74
BC040543.1 - 12 - 13 3524 74
BC051651.1 - 26 - 27 3987 74
BC151842.1 - 28 - 29 4836 74
X58288.1 - 26 - 27 3748 74

View Full Product Details