The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291902.1 | NP_001278831.1 | 14 - 15 | 3365 | 58 |
NM_002335.3 | NP_002326.2 | 14 - 15 | 3343 | 58 | |
XM_005273994.2 | XP_005274051.1 | 14 - 15 | 3338 | 58 | |
XM_011545029.1 | XP_011543331.1 | 14 - 15 | 3279 | 58 | |
XM_011545030.1 | XP_011543332.1 | 14 - 15 | 3279 | 58 | |
XM_011545031.1 | XP_011543333.1 | 14 - 15 | 3279 | 58 | |
XM_017017735.1 | XP_016873224.1 | 7 - 8 | 1559 | 58 | |
XM_017017736.1 | XP_016873225.1 | 4 - 5 | 942 | 58 | |
GenBank mRNA | AB017498.1 | - | 14 - 15 | 3340 | 58 |
AF064548.1 | - | 14 - 15 | 3285 | 58 | |
AF077820.1 | - | 14 - 15 | 3325 | 58 | |
BC098397.1 | - | 14 - 15 | 3290 | 58 | |
BC150595.1 | - | 14 - 15 | 3285 | 58 |